Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Quinolinecarboxylic acid | 86-59-9 | 173.17 | 1 ML
SDP

Catalog No. 5000460428
Encompass_Preferred

Quinoline-8-carboxylic acid is a herbicide supplied as a 10 mM solution in 1 mL of DMSO. This chemical compound has a molecular weight of 173.17 and a purity exceeding 99.7%. It is intended for research use only.

  • Supplied as a 10 mM solution in 1 mL DMSO
  • Purity >99.7%
  • Molecular weight: 173.17
  • CAS number: 86-59-9
  • Functions as a herbicide
  • Solution storage at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-60428 Supplier Medchemexpress LLC Supplier No. HYW00737110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinoline-8-carboxylic acid is a herbicide supplied as a 10 mM solution in 1 mL of DMSO. This chemical compound has a molecular weight of 173.17 and a purity exceeding 99.7%. It is intended for research use only.

  • Supplied as a 10 mM solution in 1 mL DMSO
  • Purity >99.7%
  • Molecular weight: 173.17
  • CAS number: 86-59-9
  • Functions as a herbicide
  • Solution storage at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.