Learn More
Medchemexpress LLC 8-Quinolinol, 5-chloro- | 130-16-5 | 99.9% | 500 MG
SDP
Catalog No. 5000408494
Encompass_Preferred
Cloxiquine (5-Chloro-8-quinolinol) is an antibacterial, antifungal, and antiamoebic agent. It can be used for the research of tuberculosis and dermatoses. Cloxiquine suppresses the growth and metastasis of melanoma cells through activation of PPARγ.
  • Antibacterial, antifungal, and antiamoebic agent
  • Used in research of tuberculosis and dermatoses
  • Suppresses growth and metastasis of melanoma cells
  • Activates PPARγ
  • Assayed by LCMS to 99.94% purity
Catalog No. 50-004-08494 Supplier Medchemexpress LLC Supplier No. HYB0963500MG
Cloxiquine (5-Chloro-8-quinolinol) is an antibacterial, antifungal, and antiamoebic agent. It can be used for the research of tuberculosis and dermatoses. Cloxiquine suppresses the growth and metastasis of melanoma cells through activation of PPARγ.
  • Antibacterial, antifungal, and antiamoebic agent
  • Used in research of tuberculosis and dermatoses
  • Suppresses growth and metastasis of melanoma cells
  • Activates PPARγ
  • Assayed by LCMS to 99.94% purity