Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Quinolinol, 7-[(3-ethoxy-4-methoxyphenyl)[(4-methyl-2-pyridinyl)amino]methyl]- | 430462-93-4 | 98.05% | 415.48 | 100 MG
SDP

Catalog No. 5000388123
Encompass_Preferred

HQ-415 is a biologically active metal chelator related to Clioquinol, with an EC50 of 15 μM.

  • Purity: 98.05%
  • Molecular weight: 415.48
  • Formula: C25H25N3O3
  • CAS No.: 430462-93-4
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder -20°C for 3 years, 4°C for 2 years; In solvent -80°C for 2 years, -20°C for 1 year
  • Solubility (In Vitro): DMSO : 5.6 mg/mL (13.48 mM; requires ultrasonic and warming; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Catalog No. 50-003-88123 Supplier Medchemexpress LLC Supplier No. HY18670100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HQ-415 is a biologically active metal chelator related to Clioquinol, with an EC50 of 15 μM.

  • Purity: 98.05%
  • Molecular weight: 415.48
  • Formula: C25H25N3O3
  • CAS No.: 430462-93-4
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder -20°C for 3 years, 4°C for 2 years; In solvent -80°C for 2 years, -20°C for 1 year
  • Solubility (In Vitro): DMSO : 5.6 mg/mL (13.48 mM; requires ultrasonic and warming; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.