Learn More
Medchemexpress LLC 8-Quinolinol, 7-[(3-ethoxy-4-methoxyphenyl)[(4-methyl-2-pyridinyl)amino]methyl]- | 430462-93-4 | 98.1% | 415.48 | 50 MG
SDP
Catalog No. 5000388173
Encompass_Preferred
HQ-415 is a biologically active metal chelator demonstrating an EC50 of 15 μM. This compound is structurally related to Clioquinol and appears as a solid with a white to off-white color. It is intended for research use only.
  • Biologically active.
  • Acts as a metal chelator.
  • Has an EC50 of 15 μM.
  • Provided as a solid.
Catalog No. 50-003-88173 Supplier Medchemexpress LLC Supplier No. HY1867050MG
HQ-415 is a biologically active metal chelator demonstrating an EC50 of 15 μM. This compound is structurally related to Clioquinol and appears as a solid with a white to off-white color. It is intended for research use only.
  • Biologically active.
  • Acts as a metal chelator.
  • Has an EC50 of 15 μM.
  • Provided as a solid.