Learn More
Medchemexpress LLC 8-Quinolinol, sulfate (2:1) | 134-31-6 | 99.96% | 194.20 | 1 G
SDP
Catalog No. 5000453617
Encompass_Preferred
8-Hydroxyquinoline hemisulfate is a monoprotic bidentate chelating agent. It exhibits antiseptic, disinfectant, and pesticide properties, and functions as a transcription inhibitor.
  • Monoprotic bidentate chelating agent
  • Antiseptic properties
  • Disinfectant properties
  • Pesticide properties
  • Functions as a transcription inhibitor
Catalog No. 50-004-53617 Supplier Medchemexpress LLC Supplier No. HYW0120371G
8-Hydroxyquinoline hemisulfate is a monoprotic bidentate chelating agent. It exhibits antiseptic, disinfectant, and pesticide properties, and functions as a transcription inhibitor.
  • Monoprotic bidentate chelating agent
  • Antiseptic properties
  • Disinfectant properties
  • Pesticide properties
  • Functions as a transcription inhibitor