Learn More
Integra Biosciences Corp 8-ROLL RESERVOIR 25/CS
SDP
Catalog No. NC0731319
Encompass
8 Row Reagent Reservoirs, Pyramid Bottom, Partitioned, Non-Sterile, 25 units, Polypropylene
Catalog No. NC0731319 Supplier Integra Biosciences Corp Supplier No. 6371
8 Row Reagent Reservoirs, Pyramid Bottom, Partitioned, Non-Sterile, 25 units, Polypropylene