Learn More
Arcticwhite LLC 8 ROW RESER PYRAMID BTM. 25/CS
SDP
Catalog No. NC0243302
Encompass
8 Row Reagent Reservoir, Pyramid Bottom, Polypropylene, 290mL, 0.825mL/Row, 25/Case
Catalog No. NC0243302 Supplier Arcticwhite LLC Supplier No. AWLSS30017
Add to Cart
Add to Cart
8 Row Reagent Reservoir, Pyramid Bottom, Polypropylene, 290mL, 0.825mL/Row, 25/Case