Learn More
Integra Biosciences Corp 8 ROW RESERVOIR; 25 PRE-STERIL
SDP
Catalog No. NC2114382
Encompass
8 Row Reservoir; 25 pre-sterilized standard footprint polypropylene reservoirs
This item is not returnable. View return policy
Catalog No. NC2114382 Supplier Integra Biosciences Corp Supplier No. 6372
8 Row Reservoir; 25 pre-sterilized standard footprint polypropylene reservoirs