Learn More
BRAIN RESEARCH LABORATORIES 8-SECTION STAINING NETS 64MESH
Catalog No. NC2383535
Encompass
Eight-section Staining Nets4½″ dia. x 2″ high with 64mesh nettingStaining Nets made exclusively by Brain Research Laboratories. Constructed of a clear, bonded plexiglass, permitting easy see-thru viewing of tissue.Recommended cleaning: Mild detergents only. No harsh chemicals or acids.CAUTION: Staining Nets are not suitable for xylenes or heat, not dishwasher safe.All Staining Nets are made to order and require 2-3 days before shipping.Made in the USA32mesh netting – 32 holes per inch / 64mesh netting – 64 holes per inch
Catalog No. NC2383535 Supplier BRAIN RESEARCH LABORATORIES Supplier No. 811564
Eight-section Staining Nets4½″ dia. x 2″ high with 64mesh nettingStaining Nets made exclusively by Brain Research Laboratories. Constructed of a clear, bonded plexiglass, permitting easy see-thru viewing of tissue.Recommended cleaning: Mild detergents only. No harsh chemicals or acids.CAUTION: Staining Nets are not suitable for xylenes or heat, not dishwasher safe.All Staining Nets are made to order and require 2-3 days before shipping.Made in the USA32mesh netting – 32 holes per inch / 64mesh netting – 64 holes per inch