Learn More
Medchemexpress LLC HY-N2435 5mg Medchemexpress, [8]-Shogaol CAS:36700-45-5 Purity:>98%
SDP
Catalog No. 501970392
Encompass_Preferred
Medchemexpress, HY-N2435 5mg [8]-Shogaol CAS:36700-45-5 [8]-Shogaol, one of the pungent phenolic compounds in ginger, exhibits anti-platelet activity (IC50=5 μM) and inhibits COX-2 (IC50=17.5 μM). [8]-Shogaol induces apoptosis in human leukemia cells[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0392 Supplier Medchemexpress LLC Supplier No. HYN24355MG
Medchemexpress, HY-N2435 5mg [8]-Shogaol CAS:36700-45-5 [8]-Shogaol, one of the pungent phenolic compounds in ginger, exhibits anti-platelet activity (IC50=5 μM) and inhibits COX-2 (IC50=17.5 μM). [8]-Shogaol induces apoptosis in human leukemia cells[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.