Learn More
Electron Microscopy Sciences Silicone Carbide Abrasive Film Disc, 15 µm, PSA backing
SDP
Catalog No. 50997172
Encompass_Preferred
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 15 µm
Backing: PSA
Catalog No. 50-997-172 Supplier Electron Microscopy Sciences Supplier No. 5025815
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 15 µm
Backing: PSA