Learn More
Electron Microscopy Sciences Silicone Carbide Abrasive Film Disc, 1 µm, Plain Back
SDP
Catalog No. 50997161
Encompass_Preferred
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 1 µm
Backing: Plain
Catalog No. 50-997-161 Supplier Electron Microscopy Sciences Supplier No. 502571
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 1 µm
Backing: Plain