Learn More
Electron Microscopy Sciences Silicone Carbide Abrasive Film Disc, 3 µm, Plain Back
SDP
Catalog No. 50997162
Encompass_Preferred
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 3 µm
Backing: Plain
Catalog No. 50-997-162 Supplier Electron Microscopy Sciences Supplier No. 502573
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 3 µm
Backing: Plain