Learn More
Electron Microscopy Sciences Silicone Carbide Abrasive Film Disc, 60 µm, PSA backing
SDP
Catalog No. 50997174
Encompass_Preferred
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 60 µm
Backing: PSA
Catalog No. 50-997-174 Supplier Electron Microscopy Sciences Supplier No. 5025860
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 60 µm
Backing: PSA