Learn More
Electron Microscopy Sciences Silicone Carbide Abrasive Film Disc, 9 µm, Plain Back
SDP
Catalog No. 50997164
Encompass_Preferred
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 9 µm
Backing: Plain
Catalog No. 50-997-164 Supplier Electron Microscopy Sciences Supplier No. 502579
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 9 µm
Backing: Plain