Learn More
Electron Microscopy Sciences Silicone Carbide Abrasive Film Disc, 9 µm, PSA backing
SDP
Catalog No. 50997171
Encompass_Preferred
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 9 µm
Backing: PSA
Catalog No. 50-997-171 Supplier Electron Microscopy Sciences Supplier No. 502589
Silicon carbide abrasive film discs are ready to use for polishing and grinding. Discs are 8" in diameter.
Particles: 9 µm
Backing: PSA