Learn More
Gilson Company Inc 8"SIEVE,BRASS,FULL HEIGHT,#16
Catalog No. NC2221059
Encompass_Preferred
8" Sieve, All Brass, Full Height, No. 168in (203mm) diameter ASTM E11 Test Sieve has a brass woven-wire cloth with No. 16 (1.18mm) openings and a full-height brass frame. The overall height is 2-5/8in and the stacked height is 2-1/8in.Features:Individually serial numberedCompliant with ASTM E11 requirements for Compliance SievesIncluded Items:8” Sieve, All Brass, Full Height, No. 16Certificate of Conformance to ASTM E11 requirements
This item is not returnable. View return policy
Catalog No. NC2221059 Supplier Gilson Company Inc Supplier No. V8BF #16
8" Sieve, All Brass, Full Height, No. 168in (203mm) diameter ASTM E11 Test Sieve has a brass woven-wire cloth with No. 16 (1.18mm) openings and a full-height brass frame. The overall height is 2-5/8in and the stacked height is 2-1/8in.Features:Individually serial numberedCompliant with ASTM E11 requirements for Compliance SievesIncluded Items:8” Sieve, All Brass, Full Height, No. 16Certificate of Conformance to ASTM E11 requirements