Learn More
Gilson Company Inc 8"SIEVE,S/S,FULL HEIGHT,#3-1/2
Catalog No. NC2088884
Encompass_Preferred
8" Sieve, All Stainless, Full Height, No. 3-1/2. 8in (203mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 3-1/2 (5.6mm) openings and full height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in.Features:Individually serial numberedCompliant with ASTM E11 requirements for Compliance Sieves
This item is not returnable. View return policy
Catalog No. NC2088884 Supplier Gilson Company Inc Supplier No. V8SF #31/2
Add to Cart
Add to Cart
8" Sieve, All Stainless, Full Height, No. 3-1/2. 8in (203mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 3-1/2 (5.6mm) openings and full height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in.Features:Individually serial numberedCompliant with ASTM E11 requirements for Compliance Sieves