Learn More
USA Scientific Inc 8-cap strip for 0.2 mL tubes and plates, dome top, natural
Catalog No. NC9512597
Encompass_Preferred
8-cap strip for 0.2 mL PCR tubes and plates, dome top, natural, 125 strips (1000 caps) per pack. Polypropylene.
This item is not returnable. View return policy
Catalog No. NC9512597 Supplier USA Scientific Inc Supplier No. 14000800
8-cap strip for 0.2 mL PCR tubes and plates, dome top, natural, 125 strips (1000 caps) per pack. Polypropylene.