Learn More
Chemglass Life Sciences Domed 8-Strip PCR Cap Strips, Clear, Sterile - for 0.2 mL 8-strip PCR tubes, 125/pack
SDP
Catalog No. 502733426
Encompass_Preferred
Domed 8-strip PCR cap strips provide secure, low-evaporation seals for 0.2 mL PCR strip tubes used in thermal cyclers and real-time PCR instruments. Supplied sterile in nests and molded from USP Grade VI polypropylene, they offer efficient heat transfer and reliable leak resistance for reproducible amplification results.
  • Pack size: 125 per pack.
  • Made from USP Grade VI polypropylene.
  • Compatible with most thermal cyclers and real-time PCR instruments.
  • Thin-walled design for improved thermal conductivity.
  • Domed cap design reduces liquid evaporation.
  • Easy-to-open, leak-resistant caps with reaction loss under 0.2%.
Catalog No. 50-273-3426 Supplier Chemglass Life Sciences Supplier No. CGN3610D02
Domed 8-strip PCR cap strips provide secure, low-evaporation seals for 0.2 mL PCR strip tubes used in thermal cyclers and real-time PCR instruments. Supplied sterile in nests and molded from USP Grade VI polypropylene, they offer efficient heat transfer and reliable leak resistance for reproducible amplification results.
  • Pack size: 125 per pack.
  • Made from USP Grade VI polypropylene.
  • Compatible with most thermal cyclers and real-time PCR instruments.
  • Thin-walled design for improved thermal conductivity.
  • Domed cap design reduces liquid evaporation.
  • Easy-to-open, leak-resistant caps with reaction loss under 0.2%.