Learn More
Chemglass Life Sciences Flat 8-Strip PCR Caps, Clear, Sterile, Nest - Compatible with 0.2 mL PCR 8-Strip Tubes, 125/Pack
SDP
Catalog No. 502733425
Encompass_Preferred
Chemglass CGN-3610-F-02 flat 8-strip PCR caps are clear, sterile, thin-walled polypropylene caps supplied in nests (125 per pack). They are designed for use with 0.2 mL PCR 8-strip tubes and are compatible with most thermal cyclers and real-time PCR instruments, providing efficient thermal transfer and low evaporation during amplification.
  • Made from USP Grade VI polypropylene.
  • Sterile, supplied in nests (125 per pack).
  • Thin-walled design for improved thermal conductivity.
  • Compatible with most thermal cyclers and real-time PCR instruments.
  • Low loss of reaction volume (less than 0.2%).
Catalog No. 50-273-3425 Supplier Chemglass Life Sciences Supplier No. CGN3610F02
Chemglass CGN-3610-F-02 flat 8-strip PCR caps are clear, sterile, thin-walled polypropylene caps supplied in nests (125 per pack). They are designed for use with 0.2 mL PCR 8-strip tubes and are compatible with most thermal cyclers and real-time PCR instruments, providing efficient thermal transfer and low evaporation during amplification.
  • Made from USP Grade VI polypropylene.
  • Sterile, supplied in nests (125 per pack).
  • Thin-walled design for improved thermal conductivity.
  • Compatible with most thermal cyclers and real-time PCR instruments.
  • Low loss of reaction volume (less than 0.2%).