Learn More
Paper Thermometer Co 8 TEMP THERMOLABEL 130-230F
Catalog No. NC9017033
Encompass
This item has a minimum order quantity of 12 packs per supplier requirements. 8-Temp Thermolabel. Self-adhering indicators are avail in packs of 16 labels each.
Catalog No. NC9017033 Supplier Paper Thermometer Co Supplier No. TL8130
This item has a minimum order quantity of 12 packs per supplier requirements. 8-Temp Thermolabel. Self-adhering indicators are avail in packs of 16 labels each.