Learn More
Cayman Chemical Δ8-THC methyl ether, Purity: ≥98%, CAS: 1451-20-3, 5 mg
SDP
Catalog No. 502913328
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; has antinociceptive activity in mice; intended for research and forensic applications
Catalog No. 50-291-3328 Supplier Cayman Chemical Supplier No. 367935 MG
An analytical reference standard that is structurally similar to known phytocannabinoids; has antinociceptive activity in mice; intended for research and forensic applications