Learn More
Thermco Products Inc 8 inch protective metal armor for liquid in glass thermometers
Catalog No. NC0667639
Encompass_Preferred
The metal armors are made of nickel ptated brass and have slotted front so the temperature can be easily read. A suspension ring is included to hang the thermometer
Catalog No. NC0667639 Supplier Thermco Products Inc Supplier No. ACC8ARM
The metal armors are made of nickel ptated brass and have slotted front so the temperature can be easily read. A suspension ring is included to hang the thermometer