Learn More
Chemglass Life Sciences 8" ECO Funnel, 70mm Thread, Use CG-1759-13 Waste Container
SDP
Catalog No. CG175904
Encompass_Preferred
8" ECO Funnel, 70mm Thread, Use CG-1759-13 Waste Container. The 8¿ Eco Funnel is make of HDPE and has a 2000mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.
Catalog No. CG175904 Supplier Chemglass Life Sciences Supplier No. CG175904
8" ECO Funnel, 70mm Thread, Use CG-1759-13 Waste Container. The 8¿ Eco Funnel is make of HDPE and has a 2000mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.