Learn More
Ace Glass, Inc. 8-way Tefzel tubing connector valve
SDP
Catalog No. 50977815
Encompass
8-way Tefzel tubing connector valve, for applications where multiple channels are necessary for distributing reagents, solvents or buffers to chromatography columns, etc. Valve body is machined of single-piece PTFE. Tefzel valves are rated at 500psi. The 1.5mm I.D. channels meet at the valve center with one moving part for long life/trouble free operation. Zero dead space within valve. Polypropylene caps with Fluoron elastomer O-Ring seals for instant connection of rigid or flexible tubing. 1/4" -28 thread at back of cap. Autoclaveable.
Catalog No. 50-977-815 Supplier Ace Glass, Inc. Supplier No. 1273246
8-way Tefzel tubing connector valve, for applications where multiple channels are necessary for distributing reagents, solvents or buffers to chromatography columns, etc. Valve body is machined of single-piece PTFE. Tefzel valves are rated at 500psi. The 1.5mm I.D. channels meet at the valve center with one moving part for long life/trouble free operation. Zero dead space within valve. Polypropylene caps with Fluoron elastomer O-Ring seals for instant connection of rigid or flexible tubing. 1/4" -28 thread at back of cap. Autoclaveable.