Learn More
Analytical Sales & Services 8 Well Aluminum Vial Tray for 30mm, Round Bottom Vials
Catalog No. 5000807111
Encompass
8 Well Aluminum Vial Tray for 30mm, Round Bottom Vials\n\n• SLAS footprint  (127.75mm x 85.5mm)\n\nFor use with:\n• Agilent fraction collector\n• Waters™ 2767 fraction collector\n• SP VirTis AdVantage Pro Freeze Dryer/Lyophilizer
Catalog No. 50-008-07111 Supplier Analytical Sales & Services Supplier No. 963048
8 Well Aluminum Vial Tray for 30mm, Round Bottom Vials\n\n• SLAS footprint  (127.75mm x 85.5mm)\n\nFor use with:\n• Agilent fraction collector\n• Waters™ 2767 fraction collector\n• SP VirTis AdVantage Pro Freeze Dryer/Lyophilizer