Learn More
Andwin Scientific Chamber Slide, 8 wells, 0.7 cm2 growth area per well, 0.2-0.5 ml working volume, 16/tray, 16/pack
SDP
Catalog No. 50820422
Encompass
The Lab-Tek II 8-well chamber slide system is ideal for use with viral and mycoplasma testing, chromosome studies, toxicity tests, and immunocytology. This product has 0.7 square centimeters growth area per well. It is sold in a case of 16.
Catalog No. 50-820-422 Supplier Andwin Scientific Supplier No. 46154534
The Lab-Tek II 8-well chamber slide system is ideal for use with viral and mycoplasma testing, chromosome studies, toxicity tests, and immunocytology. This product has 0.7 square centimeters growth area per well. It is sold in a case of 16.