Learn More
Cellvis 8-Well Chambered Cover Glass with Number 1.5 High Performance Cover Glass
Catalog No. NC1161532
Encompass
The 8-Well Chambered Coverglass by Cellvis is made with high quality #1.5 cover glass and has 8 wells with a volume capacity of 400-600 μL each. The chambers have individual access ports and are ideal for live cell imaging. The cover glass is compatible with most microscope objectives and optimized for high resolution imaging. The chambers are reusable, can withstand temperatures up to 80°C, and are sterilizable via autoclaving. This product enables multi-sample imaging and analysis on a single coverglass.
Catalog No. NC1161532 Supplier Cellvis Supplier No. C81.5HN
Add to Cart
Add to Cart
The 8-Well Chambered Coverglass by Cellvis is made with high quality #1.5 cover glass and has 8 wells with a volume capacity of 400-600 μL each. The chambers have individual access ports and are ideal for live cell imaging. The cover glass is compatible with most microscope objectives and optimized for high resolution imaging. The chambers are reusable, can withstand temperatures up to 80°C, and are sterilizable via autoclaving. This product enables multi-sample imaging and analysis on a single coverglass.