Learn More
Alkali Scientific 8 x 10 inch BioBlue-Lite™ Western Blot Film, 100 sheets
SDP
Catalog No. 502335986
Encompass_Preferred
Blu-Lite UHC™ is the newest line of high contrast, high sensitivity autoradiography film. Blu-Lite UHC film is specially formulated for an optimized balance of high sensitivity and low background. This low background, high contrast formula produces a lighter blue film, making it ideal for all autoradiographic and electrophoretic applications. In addition to Blu-Lite UHC’s excellent sensitivity and contrast characteristics, it is also coated on both sides (double emulsion) for improved consistency and overall performance. This new film reliably produces sharp images which allow for maximum DNA visualization with unsurpassed clarity. Compatible with automatic processors and manual development methods, Blu-Lite compares directly with films costing 3-5 times as much. With a shelf life of 2 years from date of production, Blu-Lite UHC is packaged in lined, light-safe dispenser boxes that include an improved “lift off” cover. Film is packaged 100 sheets per box.
Catalog No. 50-233-5986 Supplier Alkali Scientific Supplier No. XR8815
Blu-Lite UHC™ is the newest line of high contrast, high sensitivity autoradiography film. Blu-Lite UHC film is specially formulated for an optimized balance of high sensitivity and low background. This low background, high contrast formula produces a lighter blue film, making it ideal for all autoradiographic and electrophoretic applications. In addition to Blu-Lite UHC’s excellent sensitivity and contrast characteristics, it is also coated on both sides (double emulsion) for improved consistency and overall performance. This new film reliably produces sharp images which allow for maximum DNA visualization with unsurpassed clarity. Compatible with automatic processors and manual development methods, Blu-Lite compares directly with films costing 3-5 times as much. With a shelf life of 2 years from date of production, Blu-Lite UHC is packaged in lined, light-safe dispenser boxes that include an improved “lift off” cover. Film is packaged 100 sheets per box.