Learn More
Elkay Plastics Company Bag Gusseted Clear Poly 1 mil 8 X 4 X 18 1,000 per Case
Catalog No. NC1041995
Encompass_Preferred
Gusseted bags feature creased sides that expand to accommodate bulky items. Low Density Polyethylene (LDPE) bags are the most commonly used plastic bag because they provide excellent protection for everyday use at a great price. LKPolyethylene bags are made from virgin materials, so they comply with FDA requirements for use in food applications.
Catalog No. NC1041995 Supplier Elkay Plastics Company Supplier No. 10G084018
Gusseted bags feature creased sides that expand to accommodate bulky items. Low Density Polyethylene (LDPE) bags are the most commonly used plastic bag because they provide excellent protection for everyday use at a great price. LKPolyethylene bags are made from virgin materials, so they comply with FDA requirements for use in food applications.