Learn More
Analytical Sales & Services 8 x 40mm, 1mL Polypropylene Shell Vials with Plugs
Catalog No. 5000804486
Encompass
Advantage™ Polypropylene Shell Vials with Plugs\nSize: 8 x 40mm\nVolume: 1mL\nPlugs: 8mm, Conical, Starburst Lined\n\nQty/Pkg: 1000 (case)\n\n 
Catalog No. 50-008-04486 Supplier Analytical Sales & Services Supplier No. 30873CASE
Advantage™ Polypropylene Shell Vials with Plugs\nSize: 8 x 40mm\nVolume: 1mL\nPlugs: 8mm, Conical, Starburst Lined\n\nQty/Pkg: 1000 (case)\n\n 