Learn More
HEPATOCHEM 8 X 8 ml vials photochemistry holder for PhotoRedOx Box, Duo, and TC
Catalog No. NC1519985
Encompass_Preferred
8 X 8 ml vials photochemistry holder for PhotoRedOx Box™ Single HCK1006-01-016, PhotoRedOx Box™ Duo HCK1006-01-023, and PhotoRedOx Box™ TC HCK1006-01-025
Catalog No. NC1519985 Supplier HEPATOCHEM Supplier No. HCK100601020
8 X 8 ml vials photochemistry holder for PhotoRedOx Box™ Single HCK1006-01-016, PhotoRedOx Box™ Duo HCK1006-01-023, and PhotoRedOx Box™ TC HCK1006-01-025