Learn More
Thermo Scientific™ 8 x 8 PCR Strip Fixed Angle Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Catalog No. 75005720 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
8 x 8 PCR Strips
Catalog No. Capacity
75-005-720 8 x 8 PCR Strips
1 options
Catalog No. 75-005-720 Supplier Thermo Scientific™ Supplier No. 75005720
Add to Cart
Add to Cart
Support micro-volume protocols, such as PCR reaction set-up with this 8 x 8 PCR centrifuge rotor, capable of spinning up to 14,800rpm/ 16,800 x g.
Support micro-volume protocols, such as PCR reaction set-up, with the Thermo Scientific™ 8 x 8 PCR Strip Fixed Angle Rotor: Capable of spinning up to 15,000rpm / 17,860 x g for your PCR processing needs.
Supports micro-volume protocols, such as PCR reaction set-up, complete with our innovative Thermo Scientific technologies.
Secure, push-button Auto-Lock rotor exchange in less than 3 seconds delivers: 
  • Trouble-free rotor installation and removal
    • No tools are required
    • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory.
Certified† biocontainment rotor lid for glove-friendly, one-handed operation. 
Ordering Information:
Replacement biocontainment lid (75005730) and replacement O-rings (75005726) are sold separately. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Description 8 x 8 PCR Strip Fixed Angle Rotor
Type Fixed Angle Rotor
Capacity 8 x 8 PCR Strips
Max. RCF 17,860 x g
Max. Speed 15,000 rpm
Radius (Metric) 71 mm
K Factor 553
For Use With (Equipment) Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Angle 45°
Biocontainment Yes
† Biocontainment certification by the Public Health England, Porton Down, UK.