Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 8mm ID X 10mm OD
SDP
Catalog No. CGQ031007
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 8mm ID X 10mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031007 Supplier Chemglass Life Sciences Supplier No. CGQ031007
Graded Seal, Quartz to Borosilicate, 8mm ID X 10mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.