Learn More
Kuhner Shaker Inc 8 X TRANSPARENT POLYSTYRENE SQ
Catalog No. 502570144
Encompass
8 x transparent polystyrene square deepwell MTP (disposable, 24 wells of 11 ml volume, gamma-irradiated, with PS lid and barcode)
Catalog No. 50-257-0144 Supplier Kuhner Shaker Inc Supplier No. SMCR1424CLB
8 x transparent polystyrene square deepwell MTP (disposable, 24 wells of 11 ml volume, gamma-irradiated, with PS lid and barcode)