Learn More
Kuhner Shaker Inc 8 X TRANSPARENT POLYSTYRENE SQ
Catalog No. 502570145
Encompass
8 x transparent polystyrene square deepwell MTP (disposable, 24 wells of 11 ml volume, gamma-irradiated, lid, no notches)
Catalog No. 50-257-0145 Supplier Kuhner Shaker Inc Supplier No. SMCR1424CBMS
8 x transparent polystyrene square deepwell MTP (disposable, 24 wells of 11 ml volume, gamma-irradiated, lid, no notches)