Learn More
Kuhner Shaker Inc 8 X WHITE POLYSTYRENE SQUARE D
Catalog No. 502570141
Encompass
8 x white polystyrene square deepwell MTP (disposable, 24 wells of 11ml volume, transparant bottom, gamma irradiated)
Catalog No. 50-257-0141 Supplier Kuhner Shaker Inc Supplier No. SMCR1424E
8 x white polystyrene square deepwell MTP (disposable, 24 wells of 11ml volume, transparant bottom, gamma irradiated)