Learn More
Cayman Chemical 8(Z)-Eicosenoic Acid, Purity: ≥98%, CAS: 76261-96-6, 100 mg
SDP
Catalog No. 502986706
Encompass_Preferred
A monounsaturated fatty acid; potentiates ACh receptor channel currents without depression and enhances PCKε phosphorylation of a substrate peptide in Xenopus oocytes; constitutes 6% of the fatty acid pool in seed oil isolated from B. collina
Catalog No. 50-298-6706 Supplier Cayman Chemical Supplier No. 22579100 MG
A monounsaturated fatty acid; potentiates ACh receptor channel currents without depression and enhances PCKε phosphorylation of a substrate peptide in Xenopus oocytes; constitutes 6% of the fatty acid pool in seed oil isolated from B. collina