Learn More
Cayman Chemical 8(Z)-Eicosenoic Acid methyl ester, Purity: ≥95%, CAS: 69119-99-9, 5 mg
SDP
Catalog No. 502909796
Encompass_Preferred
An esterified form of 8(Z)-eicosenoic acid; has been used as a standard for the quantification of 8(Z)-eicosenoic acid by gas chromatography
Catalog No. 50-290-9796 Supplier Cayman Chemical Supplier No. 354285 MG
An esterified form of 8(Z)-eicosenoic acid; has been used as a standard for the quantification of 8(Z)-eicosenoic acid by gas chromatography