Learn More
Cayman Chemical 8 ZEIcosenoIc ACD methyl10mg
SDP
Catalog No. 502909831
Encompass_Preferred
An esterified form of 8(Z)-eicosenoic acid; has been used as a standard for the quantification of 8(Z)-eicosenoic acid by gas chromatography
Catalog No. 50-290-9831 Supplier Cayman Chemical Supplier No. 3542810 MG
Add to Cart
Add to Cart
An esterified form of 8(Z)-eicosenoic acid; has been used as a standard for the quantification of 8(Z)-eicosenoic acid by gas chromatography