Learn More
MSC 80/20 Inc. 45 deg T-Slotted Extrusion Support Bracket, Aluminum, Clear Anodized
Catalog No. 5000529906
Encompass
This 45-degree T-Slotted Extrusion Support Bracket is constructed from aluminum and features a clear anodized finish. With an overall length of 6 inches (160 mm), it is designed for use with 2525 series extrusions and utilizes standard T-nuts for secure fastening.
  • Designed for 45-degree T-Slotted extrusions
  • Constructed from aluminum
  • Clear anodized finish
  • Overall length of 6 inches (160 mm)
  • Compatible with 2525 series
  • Utilizes standard T-nuts
Catalog No. 50-005-29906 Supplier MSC Supplier No. 02882652
This 45-degree T-Slotted Extrusion Support Bracket is constructed from aluminum and features a clear anodized finish. With an overall length of 6 inches (160 mm), it is designed for use with 2525 series extrusions and utilizes standard T-nuts for secure fastening.
  • Designed for 45-degree T-Slotted extrusions
  • Constructed from aluminum
  • Clear anodized finish
  • Overall length of 6 inches (160 mm)
  • Compatible with 2525 series
  • Utilizes standard T-nuts