Learn More
MSC 80/20 Inc. 45 Degree Support, 15 Series, 6 in Long, Black
Catalog No. 5000511165
Encompass
This 45 degree support assembly is pre-machined with counterbores and a 45 degree miter angle on both ends. It provides a clean, flush connection and is typically used to reinforce 90 degree connections. This design is ideal for applications with heavy loads, helping to minimize beam deflection.
  • Pre-machined with counterbores and a 45 degree miter angle on both ends
  • Provides a clean, flush connection
  • Reinforces 90 degree connections, minimizing beam deflection in heavy-load applications
  • Includes all necessary hardware: socket head cap screws, washers, and T-nuts
Catalog No. 50-005-11165 Supplier MSC Supplier No. 07402498
This 45 degree support assembly is pre-machined with counterbores and a 45 degree miter angle on both ends. It provides a clean, flush connection and is typically used to reinforce 90 degree connections. This design is ideal for applications with heavy loads, helping to minimize beam deflection.
  • Pre-machined with counterbores and a 45 degree miter angle on both ends
  • Provides a clean, flush connection
  • Reinforces 90 degree connections, minimizing beam deflection in heavy-load applications
  • Includes all necessary hardware: socket head cap screws, washers, and T-nuts