Learn More
MSC 80/20 Inc. 45 Degree Support, 320 mm Long, 45 Degree Miter Angle
Catalog No. 5000528142
Encompass
This 45 degree support assembly is pre-machined with counterbores and a 45 degree miter angle on both ends. It provides a clean, flush connection, typically used to reinforce 90 degree connections. This connection method is suitable for applications with heavy loads to minimize beam deflection.
  • Assembly includes socket head cap screws, washers, and T-nuts.
  • Pre-machined with counterbores and a 45 degree miter angle on both ends.
  • Provides a clean, flush connection.
  • Reinforces 90 degree connections.
  • Minimizes beam deflection in applications with heavy loads.
Catalog No. 50-005-28142 Supplier MSC Supplier No. 32060691
This 45 degree support assembly is pre-machined with counterbores and a 45 degree miter angle on both ends. It provides a clean, flush connection, typically used to reinforce 90 degree connections. This connection method is suitable for applications with heavy loads to minimize beam deflection.
  • Assembly includes socket head cap screws, washers, and T-nuts.
  • Pre-machined with counterbores and a 45 degree miter angle on both ends.
  • Provides a clean, flush connection.
  • Reinforces 90 degree connections.
  • Minimizes beam deflection in applications with heavy loads.