Learn More
MSC 80/20 Inc. Bolt Assembly, 10-32, 0.375 in, Black Zinc
Catalog No. 5000529389
Encompass
This bolt assembly consists of a 10-32 x 0.375 inch Socket Head Cap Screw (SHCS) and a 10-32 Slide-in Economy T-Nut with a centered thread. It features a black zinc finish, providing a durable and aesthetically pleasing solution for various fastening requirements. Designed for compatibility with Series 10 and 15 profiles, this assembly ensures secure and reliable connections in your applications.
  • Comprised of a socket head cap screw and a slide-in economy t-nut
  • Thread size of 10-32 for both components
  • Socket head cap screw measures 0.375 inches in length
  • Features a black zinc finish for enhanced durability
  • Designed for use with series 10 and 15 profiles
Catalog No. 50-005-29389 Supplier MSC Supplier No. 07173362
This bolt assembly consists of a 10-32 x 0.375 inch Socket Head Cap Screw (SHCS) and a 10-32 Slide-in Economy T-Nut with a centered thread. It features a black zinc finish, providing a durable and aesthetically pleasing solution for various fastening requirements. Designed for compatibility with Series 10 and 15 profiles, this assembly ensures secure and reliable connections in your applications.
  • Comprised of a socket head cap screw and a slide-in economy t-nut
  • Thread size of 10-32 for both components
  • Socket head cap screw measures 0.375 inches in length
  • Features a black zinc finish for enhanced durability
  • Designed for use with series 10 and 15 profiles