Learn More
MSC 80/20 Inc. Double Flange Linear Bearing, Aluminum, Series 10 - 1020 Extrusion
Catalog No. 5000529452
Encompass
This double flange linear bearing is constructed from anodized aluminum, designed for linear motion applications. It is compatible with Series 10 - 1020 Extrusions.
  • Compatible with Series 10 - 1020 Extrusion.
  • Features three open T-slots.
  • Suitable for linear motion applications.
  • Designed to support hanging loads.
  • Made from aluminum.
  • Has an anodized finish.
  • Includes 3 mounting holes.
Catalog No. 50-005-29452 Supplier MSC Supplier No. 07173172
This double flange linear bearing is constructed from anodized aluminum, designed for linear motion applications. It is compatible with Series 10 - 1020 Extrusions.
  • Compatible with Series 10 - 1020 Extrusion.
  • Features three open T-slots.
  • Suitable for linear motion applications.
  • Designed to support hanging loads.
  • Made from aluminum.
  • Has an anodized finish.
  • Includes 3 mounting holes.