Learn More
MSC 80/20 Inc. Dual Roller Bearing, Nylon, Series 40, M8x1.25
Catalog No. 5000512833
Encompass
This dual roller bearing is engineered for use with Series 40 profiles. It is constructed from durable nylon, ensuring reliable performance, and features an M8x1.25 thread size for secure integration.
  • Constructed from nylon
  • Designed for use with Series 40 profiles
  • Features an M8x1.25 thread size
  • Available in black color
Catalog No. 50-005-12833 Supplier MSC Supplier No. 54969605
This dual roller bearing is engineered for use with Series 40 profiles. It is constructed from durable nylon, ensuring reliable performance, and features an M8x1.25 thread size for secure integration.
  • Constructed from nylon
  • Designed for use with Series 40 profiles
  • Features an M8x1.25 thread size
  • Available in black color