Learn More
MSC 80/20 Inc. Fastening Assembly, Series 20, Standard T-Nut, M5x0.8, Bright Zinc
Catalog No. 5000514290
Encompass
This fastening assembly is designed for use with Series 20 extrusions, providing a secure connection for various applications. It includes a standard T-nut and a socket head cap screw, featuring an M5x0.8 thread size and a durable bright zinc finish.
  • Designed for use with Series 20 extrusions.
  • Includes a standard T-nut.
  • Features a socket head cap screw.
  • M5x0.8 thread size for secure fastening.
  • Bright zinc finish for enhanced durability.
Catalog No. 50-005-14290 Supplier MSC Supplier No. 81328700
This fastening assembly is designed for use with Series 20 extrusions, providing a secure connection for various applications. It includes a standard T-nut and a socket head cap screw, featuring an M5x0.8 thread size and a durable bright zinc finish.
  • Designed for use with Series 20 extrusions.
  • Includes a standard T-nut.
  • Features a socket head cap screw.
  • M5x0.8 thread size for secure fastening.
  • Bright zinc finish for enhanced durability.