Learn More
MSC 80/20 Inc. Framing Upright, 72 in, Anodized Aluminum
Catalog No. 5000534987
Encompass
This framing upright is manufactured by 80/20 Inc. and designed for various framing applications. It features a sturdy construction with a height of 72 inches and is made from durable anodized aluminum.
  • Made from aluminum
  • Overall height of 72 in
  • Features an anodized finish
  • Clear color
Catalog No. 50-005-34987 Supplier MSC Supplier No. 61984944
This framing upright is manufactured by 80/20 Inc. and designed for various framing applications. It features a sturdy construction with a height of 72 inches and is made from durable anodized aluminum.
  • Made from aluminum
  • Overall height of 72 in
  • Features an anodized finish
  • Clear color