Learn More
MSC 80/20 Inc. Hidden Corner Connector, Inside-Inside, Zinc
Catalog No. 5000512041
Encompass
This hidden corner connector provides a 90-degree connection, allowing for discrete assembly. It includes set screws for quick and easy installation, suitable for connecting the inside of two profiles or between the inside of one and the outside of another. This connector offers a cost-effective fastening solution as it does not require additional machining.
  • Provides a 90-degree hidden corner connection.
  • Includes set screws for quick and easy connections.
  • Can connect between the inside of two profiles or inside of one and outside of another.
  • Requires no machining for installation.
  • Offers a cost-effective fastening option.
  • Recommended tool for hardware is a 3mm hex wrench.
Catalog No. 50-005-12041 Supplier MSC Supplier No. 62959218
This hidden corner connector provides a 90-degree connection, allowing for discrete assembly. It includes set screws for quick and easy installation, suitable for connecting the inside of two profiles or between the inside of one and the outside of another. This connector offers a cost-effective fastening solution as it does not require additional machining.
  • Provides a 90-degree hidden corner connection.
  • Includes set screws for quick and easy connections.
  • Can connect between the inside of two profiles or inside of one and outside of another.
  • Requires no machining for installation.
  • Offers a cost-effective fastening option.
  • Recommended tool for hardware is a 3mm hex wrench.